missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-2-[Hydroxy(diphenyl)methyl]-1-methylpyrrolidine 98.0+%, TCI America™

Catalog No. p-7130720
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H07841G 1 g
H07845G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H07841G Supplier TCI America Supplier No. H07841G
Only null left

Chemical Identifiers

CAS 144119-12-0
Molecular Formula C18H22NO
Molecular Weight (g/mol) 268.38
MDL Number MFCD00145217
InChI Key XIJAGFLYYNXCAB-QGZVFWFLSA-O
Synonym r-1-methylpyrrolidin-2-yl diphenylmethanol, r---2-hydroxy diphenyl methyl-1-methylpyrrolidine, r-alpha,alpha-diphenylmethylprolinol, 2r-1-methylpyrrolidin-2-yl-diphenylmethanol, 1,1-diphenyl-n-methyl-d-prolinol, 2r-1-methylpyrrolidin-2-yl diphenylmethanol, r-n-methyl-alpha,alpha-diphenyl-2-pyrrolidinemethanol, r-a,a-diphenylmethylprolinol, d-diphenylmethylprolinol, r---diphenyl 1-methylpyrrolidin-2-yl methanol
PubChem CID 853010
IUPAC Name (2R)-2-(hydroxydiphenylmethyl)-1-methylpyrrolidin-1-ium
SMILES C[NH+]1CCC[C@@H]1C(O)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 72°C
Color White-Yellow
Quantity 1 g
Formula Weight 267.37
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material (R)-(-)-2-[Hydroxy(diphenyl)methyl]-1-methylpyrrolidine