accessibility menu, dialog, popup

UserName

CH3NH(R)OH

CAS RN 144119-12-0

IUPAC Name: (2R)-2-(hydroxydiphenylmethyl)-1-methylpyrrolidin-1-ium
Synonyms: (2R)-2-(hydroxydiphenylmethyl)-1-methylpyrrolidin-1-ium
Molecular Weight (g/mol): 268.38
Molecular Formula: C18H22NO
InChi Key: XIJAGFLYYNXCAB-QGZVFWFLSA-O
SMILES: C[NH+]1CCC[C@@H]1C(O)(C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results