missing translation for 'onlineSavingsMsg'
Learn More

(R)-2-(Fmoc-amino)butyric acid, 98%

CAS: 170642-27-0 | C19H19NO4 | 325.364 g/mol

$64.44 - $743.19

Chemical Identifiers

CAS 170642-27-0
Molecular Formula C19H19NO4
Molecular Weight (g/mol) 325.364
MDL Number MFCD00270336
InChI Key XQIRYUNKLVPVRR-QGZVFWFLSA-N
Synonym fmoc-d-abu-oh, r-2-9h-fluoren-9-yl methoxy carbonyl amino butanoic acid, 2r-2-9h-fluoren-9-ylmethoxy carbonyl amino butanoic acid, 2r-2-9h-fluoren-9-ylmethoxycarbonylamino butanoic acid, butanoic acid, 2-9h-fluoren-9-ylmethoxy carbonyl amino-, 2r, l-2-fmoc-amino butyric acid, fmoc-d-2-aminobutyric acid, fmoc-d-abu 2-oh, ambotzfaa1611, fmoc-d-2abu-oh
PubChem CID 853017
IUPAC Name (2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)butanoic acid
SMILES CCC(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6392303
View Documents
Thermo Scientific Chemicals
H6392303
1 g
Each for $64.44
Only null left
 
AAH6392306
View Documents
Thermo Scientific Chemicals
H6392306
5 g
Each for $201.25
Only null left
 
AAH6392314
View Documents
Thermo Scientific Chemicals
H6392314
25 g
Each for $743.19
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

170642-27-0
325.364
XQIRYUNKLVPVRR-QGZVFWFLSA-N
853017
CCC(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13
C19H19NO4
MFCD00270336
fmoc-d-abu-oh, r-2-9h-fluoren-9-yl methoxy carbonyl amino butanoic acid, 2r-2-9h-fluoren-9-ylmethoxy carbonyl amino butanoic acid, 2r-2-9h-fluoren-9-ylmethoxycarbonylamino butanoic acid, butanoic acid, 2-9h-fluoren-9-ylmethoxy carbonyl amino-, 2r, l-2-fmoc-amino butyric acid, fmoc-d-2-aminobutyric acid, fmoc-d-abu 2-oh, ambotzfaa1611, fmoc-d-2abu-oh
(2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)butanoic acid

Specifications

170642-27-0
MFCD00270336
fmoc-d-abu-oh, r-2-9h-fluoren-9-yl methoxy carbonyl amino butanoic acid, 2r-2-9h-fluoren-9-ylmethoxy carbonyl amino butanoic acid, 2r-2-9h-fluoren-9-ylmethoxycarbonylamino butanoic acid, butanoic acid, 2-9h-fluoren-9-ylmethoxy carbonyl amino-, 2r, l-2-fmoc-amino butyric acid, fmoc-d-2-aminobutyric acid, fmoc-d-abu 2-oh, ambotzfaa1611, fmoc-d-2abu-oh
CCC(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13
325.364
325.36
Solid
C19H19NO4
1 g
XQIRYUNKLVPVRR-QGZVFWFLSA-N
(2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)butanoic acid
853017
98%
(R)-2-(Fmoc-amino)butyric acid
Videos
Safety and Handling

Safety and Handling

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only