missing translation for 'onlineSavingsMsg'
Learn More

(R)-2-(Fmoc-amino)butyric acid, 98%

CAS: 170642-27-0 | C19H19NO4 | 325.364 g/mol

Supplier:  Thermo Scientific Chemicals H6392306

 View more versions of this product

Catalog No. AAH6392306


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

(R)-2-(Fmoc-amino)butyric acid
C19H19NO4
5 g
XQIRYUNKLVPVRR-QGZVFWFLSA-N
(2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)butanoic acid
853017
98%
170642-27-0
MFCD00270336
fmoc-d-abu-oh, r-2-9h-fluoren-9-yl methoxy carbonyl amino butanoic acid, 2r-2-9h-fluoren-9-ylmethoxy carbonyl amino butanoic acid, 2r-2-9h-fluoren-9-ylmethoxycarbonylamino butanoic acid, butanoic acid, 2-9h-fluoren-9-ylmethoxy carbonyl amino-, 2r, l-2-fmoc-amino butyric acid, fmoc-d-2-aminobutyric acid, fmoc-d-abu 2-oh, ambotzfaa1611, fmoc-d-2abu-oh
CCC(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13
325.364
325.36
Solid

Chemical Identifiers

170642-27-0
325.364
XQIRYUNKLVPVRR-QGZVFWFLSA-N
853017
CCC(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13
C19H19NO4
MFCD00270336
fmoc-d-abu-oh, r-2-9h-fluoren-9-yl methoxy carbonyl amino butanoic acid, 2r-2-9h-fluoren-9-ylmethoxy carbonyl amino butanoic acid, 2r-2-9h-fluoren-9-ylmethoxycarbonylamino butanoic acid, butanoic acid, 2-9h-fluoren-9-ylmethoxy carbonyl amino-, 2r, l-2-fmoc-amino butyric acid, fmoc-d-2-aminobutyric acid, fmoc-d-abu 2-oh, ambotzfaa1611, fmoc-d-2abu-oh
(2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)butanoic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only