missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(R)-(+)-2-Diphenylphosphino-2'-methoxy-1,1'-binaphthyl 98.0+%, TCI America™
$292.62 - $1418.85
Chemical Identifiers
| CAS | 145964-33-6 |
|---|---|
| Molecular Formula | C33H25OP |
| Molecular Weight (g/mol) | 468.536 |
| MDL Number | MFCD00269686 |
| InChI Key | KRWTWSSMURUMDE-UHFFFAOYSA-N |
| Synonym | (R)-(+)-MOP |
| PubChem CID | 4190002 |
| IUPAC Name | [1-(2-methoxynaphthalen-1-yl)naphthalen-2-yl]-diphenylphosphane |
| SMILES | COC1=C(C2=CC=CC=C2C=C1)C3=C(C=CC4=CC=CC=C43)P(C5=CC=CC=C5)C6=CC=CC=C6 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
D2774100MG
|
TCI America
D2774100MG |
100 mg |
Each for $292.62
|
|
|||||
|
D27741G
|
TCI America
D27741G |
1 g |
Each for $1,418.85
|
|
|||||
Chemical Identifiers
| 145964-33-6 | |
| 468.536 | |
| KRWTWSSMURUMDE-UHFFFAOYSA-N | |
| 4190002 | |
| COC1=C(C2=CC=CC=C2C=C1)C3=C(C=CC4=CC=CC=C43)P(C5=CC=CC=C5)C6=CC=CC=C6 |
| C33H25OP | |
| MFCD00269686 | |
| (R)-(+)-MOP | |
| [1-(2-methoxynaphthalen-1-yl)naphthalen-2-yl]-diphenylphosphane |
Specifications
| 145964-33-6 | |
| White-Yellow | |
| MFCD00269686 | |
| (R)-(+)-MOP | |
| COC1=C(C2=CC=CC=C2C=C1)C3=C(C=CC4=CC=CC=C43)P(C5=CC=CC=C5)C6=CC=CC=C6 | |
| 468.536 | |
| 468.54 | |
| Crystalline Powder |
| 193°C | |
| C33H25OP | |
| 100 mg | |
| KRWTWSSMURUMDE-UHFFFAOYSA-N | |
| [1-(2-methoxynaphthalen-1-yl)naphthalen-2-yl]-diphenylphosphane | |
| 4190002 | |
| ≥98.0% (GC) | |
| (R)-(+)-2-Diphenylphosphino-2′-methoxy-1,1′-binaphthyl |
Safety and Handling
TSCA : No