missing translation for 'onlineSavingsMsg'
Learn More

(R)-(+)-2-Diphenylphosphino-2'-methoxy-1,1'-binaphthyl 98.0+%, TCI America™

Catalog No. d2774100mg
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
100 mg
Catalog No. Quantity
D2774100MG 100 mg
D27741G 1 g
2 options

Catalog No. D2774100MG

Supplier: TCI America D2774100MG

Only null left

Chemical Identifiers

CAS 145964-33-6
Molecular Formula C33H25OP
Molecular Weight (g/mol) 468.536
MDL Number MFCD00269686
InChI Key KRWTWSSMURUMDE-UHFFFAOYSA-N
Synonym (R)-(+)-MOP
PubChem CID 4190002
IUPAC Name [1-(2-methoxynaphthalen-1-yl)naphthalen-2-yl]-diphenylphosphane
SMILES COC1=C(C2=CC=CC=C2C=C1)C3=C(C=CC4=CC=CC=C43)P(C5=CC=CC=C5)C6=CC=CC=C6

Specifications

Melting Point 193°C
Color White-Yellow
Quantity 100 mg
Formula Weight 468.54
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material (R)-(+)-2-Diphenylphosphino-2′-methoxy-1,1′-binaphthyl