missing translation for 'onlineSavingsMsg'
Learn More

(R)-(+)-2-Amino-2'-hydroxy-1,1'-binaphthyl 98.0+%, TCI America™

Catalog No. A2316100MG
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A2316100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A2316100MG Supplier TCI America Supplier No. A2316100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 137848-28-3
Molecular Formula C20H15NO
Molecular Weight (g/mol) 285.346
MDL Number MFCD01882346
InChI Key HIXQCPGXQVQHJP-UHFFFAOYSA-N
Synonym (R)-(+)-1-(2-Amino-1-naphthyl)-2-naphthol, (R)-(+)-NOBIN, (R)-(+)-2′C-Amino-1,1′C-binaphthalen-2-ol
PubChem CID 3617797
IUPAC Name 1-(2-aminonaphthalen-1-yl)naphthalen-2-ol
SMILES C1=CC=C2C(=C1)C=CC(=C2C3=C(C=CC4=CC=CC=C43)O)N

Specifications

Melting Point 173°C
Quantity 100 mg
Formula Weight 285.35
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material (R)-(+)-2-Amino-2′-hydroxy-1,1′-binaphthyl