accessibility menu, dialog, popup

UserName

CAS RN 137848-28-3

IUPAC Name: 2'-amino-[1,1'-binaphthalen]-2-ol
Synonyms: nobin
Molecular Weight (g/mol): 285.35
Molecular Formula: C20H15NO
InChi Key: HIXQCPGXQVQHJP-UHFFFAOYSA-N
SMILES: NC1=C(C2=CC=CC=C2C=C1)C1=C2C=CC=CC2=CC=C1O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results