Learn More
(R)-(+)-2,2'-Bis(di-p-tolylphosphino)-1,1'-binaphthyl, 98%
Chiral catalyst ligand | CAS: 99646-28-3 | C48H40P2 | 678.796 g/mol
Supplier: Thermo Scientific Chemicals 04211902
Description
(R)-(+)-2,2'-Bis(di-p-tolylphosphino)-1,1'-binaphthyl is used as chiral catalyst ligand. (R)-T-BINAP complexes derived from rhodium precursors are used for the asymmetric hydroformylation of vinyl acetate. It is a catalysts used for reductive amination of ketones, Pt dications for cation trapping, Rh(I)-catalyst for hydrogenation of acetamidoacrylic acid derivatives.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Spécifications
| (R)-(+)-2,2'-Bis(di-p-tolylphosphino)-1,1'-binaphthyl | |
| 255°C to 257°C | |
| C48H40P2 | |
| 0.5 g | |
| Insoluble in water. | |
| +156°C (c=0.5 in Benzene) | |
| [1-[2-bis(4-methylphenyl)phosphanylnaphthalen-1-yl]naphthalen-2-yl]-bis(4-methylphenyl)phosphane | |
| 4189906 | |
| 98% |
| 99646-28-3 | |
| White | |
| MFCD00269856 | |
| r-tol-binap, s-tol-binap, s---2,2'-bis di-p-tolylphosphino-1,1'-binaphthyl, 2,2'-bis di-p-tolylphosphino-1,1'-binaphthyl, s-t-binap, rac-tol-binap, r-+-2,2'-bis di-p-tolylphosphino-1,1'-binaphthyl, tol-binap, s-2,2'-bis di-p-tolylphosphino-1,1'-binaphthyl, s-p-tol-binap | |
| IOPQYDKQISFMJI-UHFFFAOYSA-N | |
| CC1=CC=C(C=C1)P(C2=CC=C(C=C2)C)C3=C(C4=CC=CC=C4C=C3)C5=C(C=CC6=CC=CC=C65)P(C7=CC=C(C=C7)C)C8=CC=C(C=C8)C | |
| 678.796 | |
| 678.79 | |
| Powder |
Identifiants chimiques
| 99646-28-3 | |
| 678.796 | |
| IOPQYDKQISFMJI-UHFFFAOYSA-N | |
| 4189906 | |
| CC1=CC=C(C=C1)P(C2=CC=C(C=C2)C)C3=C(C4=CC=CC=C4C=C3)C5=C(C=CC6=CC=CC=C65)P(C7=CC=C(C=C7)C)C8=CC=C(C=C8)C |
| C48H40P2 | |
| MFCD00269856 | |
| r-tol-binap, s-tol-binap, s---2,2'-bis di-p-tolylphosphino-1,1'-binaphthyl, 2,2'-bis di-p-tolylphosphino-1,1'-binaphthyl, s-t-binap, rac-tol-binap, r-+-2,2'-bis di-p-tolylphosphino-1,1'-binaphthyl, tol-binap, s-2,2'-bis di-p-tolylphosphino-1,1'-binaphthyl, s-p-tol-binap | |
| [1-[2-bis(4-methylphenyl)phosphanylnaphthalen-1-yl]naphthalen-2-yl]-bis(4-methylphenyl)phosphane |
Sécurité et manipulation
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only