missing translation for 'onlineSavingsMsg'
Learn More

(R)-2,2'-Bis[bis(4-methoxy-3,5-dimethylphenyl)phosphino]-4,4',6,6'-tetramethoxybiphenyl, 97+%

CAS: 1365531-93-6 | C52H60O8P2 | 874.992 g/mol

$284.22 - $1004.45

Chemical Identifiers

CAS 1365531-93-6
Molecular Formula C52H60O8P2
Molecular Weight (g/mol) 874.992
MDL Number MFCD19443626
InChI Key PKHREKHNGLJKHV-UHFFFAOYSA-N
Synonym 2'-bis 4-methoxy-3,5-dimethylphenyl phosphanyl-4,4',6,6'-tetramethoxy-1,1'-biphenyl-2-yl bis 4-methoxy-3,5-dimethylphenyl phosphane, ar-2,2',4,4'-tetramethoxy-6,6'-bis bis 3,5-dimethyl-4-methoxyphenyl phosphino biphenyl, r-2,2'-bis bis 4-methoxy-3,5-dimethylphenyl phosphino-4,4',6,6'-tetramethoxybiphenyl, s-2,2'-bis bis 4-methoxy-3,5-dimethylphenyl phosphino-4,4',6,6'-tetramethoxybiphenyl, r-4,4',6,6'-tetramethoxybiphenyl-2,2'-diyl bis bis 4-methoxy-3,5-dimethylphenyl phosphine, s-4,4',6,6'-tetramethoxybiphenyl-2,2'-diyl bis bis 4-methoxy-3,5-dimethylphenyl phosphine
PubChem CID 87110691
IUPAC Name [2-[2-bis(4-methoxy-3,5-dimethylphenyl)phosphanyl-4,6-dimethoxyphenyl]-3,5-dimethoxyphenyl]-bis(4-methoxy-3,5-dimethylphenyl)phosphane
SMILES CC1=CC(=CC(=C1OC)C)P(C2=CC(=C(C(=C2)C)OC)C)C3=CC(=CC(=C3C4=C(C=C(C=C4P(C5=CC(=C(C(=C5)C)OC)C)C6=CC(=C(C(=C6)C)OC)C)OC)OC)OC)OC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH60684MD
View Documents
Thermo Scientific Chemicals
H60684MD
250 mg
Each for $284.22
Only null left
 
AAH6068403
View Documents
Thermo Scientific Chemicals
H6068403
1 g
Each for $1,004.45
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1365531-93-6
874.992
PKHREKHNGLJKHV-UHFFFAOYSA-N
87110691
CC1=CC(=CC(=C1OC)C)P(C2=CC(=C(C(=C2)C)OC)C)C3=CC(=CC(=C3C4=C(C=C(C=C4P(C5=CC(=C(C(=C5)C)OC)C)C6=CC(=C(C(=C6)C)OC)C)OC)OC)OC)OC
C52H60O8P2
MFCD19443626
2'-bis 4-methoxy-3,5-dimethylphenyl phosphanyl-4,4',6,6'-tetramethoxy-1,1'-biphenyl-2-yl bis 4-methoxy-3,5-dimethylphenyl phosphane, ar-2,2',4,4'-tetramethoxy-6,6'-bis bis 3,5-dimethyl-4-methoxyphenyl phosphino biphenyl, r-2,2'-bis bis 4-methoxy-3,5-dimethylphenyl phosphino-4,4',6,6'-tetramethoxybiphenyl, s-2,2'-bis bis 4-methoxy-3,5-dimethylphenyl phosphino-4,4',6,6'-tetramethoxybiphenyl, r-4,4',6,6'-tetramethoxybiphenyl-2,2'-diyl bis bis 4-methoxy-3,5-dimethylphenyl phosphine, s-4,4',6,6'-tetramethoxybiphenyl-2,2'-diyl bis bis 4-methoxy-3,5-dimethylphenyl phosphine
[2-[2-bis(4-methoxy-3,5-dimethylphenyl)phosphanyl-4,6-dimethoxyphenyl]-3,5-dimethoxyphenyl]-bis(4-methoxy-3,5-dimethylphenyl)phosphane

Specifications

1365531-93-6
MFCD19443626
Air Sensitive
PKHREKHNGLJKHV-UHFFFAOYSA-N
[2-[2-bis(4-methoxy-3,5-dimethylphenyl)phosphanyl-4,6-dimethoxyphenyl]-3,5-dimethoxyphenyl]-bis(4-methoxy-3,5-dimethylphenyl)phosphane
87110691
≥97%
C52H60O8P2
250 mg
2'-bis 4-methoxy-3,5-dimethylphenyl phosphanyl-4,4',6,6'-tetramethoxy-1,1'-biphenyl-2-yl bis 4-methoxy-3,5-dimethylphenyl phosphane, ar-2,2',4,4'-tetramethoxy-6,6'-bis bis 3,5-dimethyl-4-methoxyphenyl phosphino biphenyl, r-2,2'-bis bis 4-methoxy-3,5-dimethylphenyl phosphino-4,4',6,6'-tetramethoxybiphenyl, s-2,2'-bis bis 4-methoxy-3,5-dimethylphenyl phosphino-4,4',6,6'-tetramethoxybiphenyl, r-4,4',6,6'-tetramethoxybiphenyl-2,2'-diyl bis bis 4-methoxy-3,5-dimethylphenyl phosphine, s-4,4',6,6'-tetramethoxybiphenyl-2,2'-diyl bis bis 4-methoxy-3,5-dimethylphenyl phosphine
CC1=CC(=CC(=C1OC)C)P(C2=CC(=C(C(=C2)C)OC)C)C3=CC(=CC(=C3C4=C(C=C(C=C4P(C5=CC(=C(C(=C5)C)OC)C)C6=CC(=C(C(=C6)C)OC)C)OC)OC)OC)OC
874.992
874.98
(R)-2,2'-Bis[bis(4-methoxy-3,5-dimethylphenyl)phosphino]-4,4',6,6'-tetramethoxybiphenyl
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only