accessibility menu, dialog, popup

UserName

CH3OH3CH3CPH3COCH3CH3PCH3OCH3CH3CH3OCH3H3COH3COH3COCH3OCH3(?)

CAS RN 1365531-93-6

IUPAC Name: {2'-[bis(4-methoxy-3,5-dimethylphenyl)phosphanyl]-4,4',6,6'-tetramethoxy-[1,1'-biphenyl]-2-yl}bis(4-methoxy-3,5-dimethylphenyl)phosphane
Synonyms: {2'-[bis(4-methoxy-3,5-dimethylphenyl)phosphanyl]-4,4',6,6'-tetramethoxy-[1,1'-biphenyl]-2-yl}bis(4-methoxy-3,5-dimethylphenyl)phosphane
Molecular Weight (g/mol): 874.99
Molecular Formula: C52H60O8P2
InChi Key: PKHREKHNGLJKHV-UHFFFAOYSA-N
SMILES: COC1=CC(OC)=C(C(=C1)P(C1=CC(C)=C(OC)C(C)=C1)C1=CC(C)=C(OC)C(C)=C1)C1=C(OC)C=C(OC)C=C1P(C1=CC(C)=C(OC)C(C)=C1)C1=CC(C)=C(OC)C(C)=C1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results