missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-1-Benzyl-3-aminopyrrolidine, 99%, ee 99%

Catalog No. p-7047734
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1969003 1 g
AAL1969006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1969003 Supplier Thermo Scientific Chemicals Supplier No. L1969003

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 114715-39-8 | C11H16N2 | 176.26 g/mol

(R)-(-)-1-Benzyl-3-aminopyrrolidine is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(R)-(-)-1-Benzyl-3-aminopyrrolidine is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

Solubility
Slightly soluble in water.

Notes
Air sensitive. Store under inert gas. Store away from oxidizing agents, air, carbondioxide.

Chemical Identifiers

CAS 114715-39-8
Molecular Formula C11H16N2
Molecular Weight (g/mol) 176.26
MDL Number MFCD00082638
InChI Key HBVNLKQGRZPGRP-LLVKDONJSA-N
Synonym r---1-benzyl-3-aminopyrrolidine, r-1-benzyl-3-aminopyrrolidine, r-1-benzylpyrrolidin-3-amine, 3r---1-benzyl-3-aminopyrrolidine, 3r-1-benzylpyrrolidin-3-amine, r-3-amino-1-n-benzyl-pyrrolidine, r-n-benzyl-3-aminopyrrolidine, 3-pyrrolidinamine, 1-phenylmethyl-, 3r, r-3-amino-1-benzylpyrrolidine, 3r-1-benzyl-3-pyrrolidinamine
PubChem CID 1519354
IUPAC Name (3R)-1-benzylpyrrolidin-3-amine
SMILES N[C@@H]1CCN(CC2=CC=CC=C2)C1

Specifications

Density 1.02
Boiling Point 101°C to 103°C (3 mmHg)
Flash Point 140°C (284°F)
Refractive Index 1.543
Quantity 1 g
UN Number UN2735
Beilstein 6477592
Sensitivity Air Sensitive
Solubility Information Slightly soluble in water.
Optical Rotation −2° (Neat)
Formula Weight 176.26
Percent Purity 99%
Assay ee 99%
Chemical Name or Material (R)-(-)-1-Benzyl-3-aminopyrrolidine
Show More Show Less
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only