missing translation for 'onlineSavingsMsg'
Learn More

(R)-1-[(9H-Fluoren-9-ylmethoxy)carbonyl]-2-piperidinecarboxylic Acid 98.0+%, TCI America™

Catalog No. F09371G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F09371G 1 g
F09375G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F09371G Supplier TCI America Supplier No. F09371G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 101555-63-9
Molecular Formula C21H21NO4
Molecular Weight (g/mol) 351.402
MDL Number MFCD00235899
InChI Key CKLAZLINARHOTG-LJQANCHMSA-N
Synonym fmoc-d-pipecolic acid, fmoc-d-pip-oh, fmoc-d-hopro-oh, d-1-fmoc-pipecolinic acid, fmoc-d-piperidine-2-carboxylic acid, fmoc-d-homopro-oh, d-n-fmoc-pipecolic acid, r-n-fmoc-piperidine-2-carboxylic acid, r-1-9h-fluoren-9-yl methoxy carbonyl piperidine-2-carboxylic acid, fmoc-d-homoproline
PubChem CID 6958378
IUPAC Name (2R)-1-(9H-fluoren-9-ylmethoxycarbonyl)piperidine-2-carboxylic acid
SMILES C1CCN(C(C1)C(=O)O)C(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24

Specifications

Melting Point 156°C
Color White
Quantity 1 g
Formula Weight 351.40
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material (R)-1-[(9H-Fluoren-9-ylmethoxy)carbonyl]-2-piperidinecarboxylic Acid
TSCA No
Recommended Storage Refrigerator