accessibility menu, dialog, popup

UserName

CAS RN 101555-63-9

IUPAC Name: 1-{[(9H-fluoren-9-yl)methoxy]carbonyl}piperidine-2-carboxylic acid
Synonyms: 1-[(9H-fluoren-9-ylmethoxy)carbonyl]piperidine-2-carboxylic acid
Molecular Weight (g/mol): 351.4
Molecular Formula: C21H21NO4
InChi Key: CKLAZLINARHOTG-UHFFFAOYNA-N
SMILES: OC(=O)C1CCCCN1C(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results