missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(R)-(-)-1,2,3,4-Tetrahydro-1-naphthol 98.0+%, TCI America™
Supplier: TCI America T2359100MG
Specifications
| (R)-(-)-1,2,3,4-Tetrahydro-1-naphthol | |
| 40°C | |
| 140°C | |
| MFCD00063005 | |
| (R)-(-)-alpha-Tetralol | |
| C1CC(C2=CC=CC=C2C1)O | |
| 148.205 | |
| 148.21 | |
| Crystalline Powder |
| 23357-45-1 | |
| White-Yellow | |
| C10H12O | |
| 100 mg | |
| JAAJQSRLGAYGKZ-SNVBAGLBSA-N | |
| (1R)-1,2,3,4-tetrahydronaphthalen-1-ol | |
| 7058028 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 23357-45-1 | |
| 148.205 | |
| JAAJQSRLGAYGKZ-SNVBAGLBSA-N | |
| 7058028 | |
| C1CC(C2=CC=CC=C2C1)O |
| C10H12O | |
| MFCD00063005 | |
| (R)-(-)-alpha-Tetralol | |
| (1R)-1,2,3,4-tetrahydronaphthalen-1-ol |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator