accessibility menu, dialog, popup

UserName

CAS RN 23357-45-1

OH

CAS RN 23357-45-1

IUPAC Name: 1,2,3,4-tetrahydronaphthalen-1-ol
Synonyms: tetrahydronaphthalenol
Molecular Weight (g/mol): 148.21
Molecular Formula: C10H12O
InChi Key: JAAJQSRLGAYGKZ-UHFFFAOYNA-N
SMILES: OC1CCCC2=CC=CC=C12
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results