missing translation for 'onlineSavingsMsg'
Learn More

(R)-1,2,3,4-Tetrahydro-1-naphthoic acid, 98%

CAS: 23357-47-3 | C11H12O2 | 176.215 g/mol

$130.27 - $1064.48

Chemical Identifiers

CAS 23357-47-3
Molecular Formula C11H12O2
Molecular Weight (g/mol) 176.215
MDL Number MFCD07368364
InChI Key VDLWTJCSPSUGOA-SNVBAGLBSA-N
Synonym r-1,2,3,4-tetrahedro-naphthoic acid, r-1,2,3,4-tetrahydronaphthoic acid, r-1,2,3,4-tetrahydro-naphthoic acid, r-1,2,3,4-tetrahydronaphthalene-1-carboxylic acid, r-1,2,3,4-tetrahydro-1-naphthoic acid, r-1,2,3,4-tetrahedronaphthoic acid, 1r-1,2,3,4-tetrahydronaphthalene-1-carboxylic acid, 1-naphthalenecarboxylicacid, 1,2,3,4-tetrahydro-, 1r, pubchem5734, r-1,2,3,4-tert-ahydro naphthoic acid
PubChem CID 12598147
IUPAC Name (1R)-1,2,3,4-tetrahydronaphthalene-1-carboxylic acid
SMILES C1CC(C2=CC=CC=C2C1)C(=O)O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH27673MD
View Documents
Thermo Scientific Chemicals
H27673MD
250 mg
Each for $130.27
Only null left
 
AAH2767303
View Documents
Thermo Scientific Chemicals
H2767303
1 g
Each for $315.99
Only null left
 
AAH2767306
View Documents
Thermo Scientific Chemicals
H2767306
5 g
Each for $1,064.48
Only null left
 
Description

Description

It is a palonosetron intermediate. Tetrahydronaphthoic acid is used for the construction of sterically constrained protein turn models.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a palonosetron intermediate. Tetrahydronaphthoic acid is used for the construction of sterically constrained protein turn models.

Solubility
Slightly soluble in water.

Notes
Store in cool dry conditions. Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

23357-47-3
176.215
VDLWTJCSPSUGOA-SNVBAGLBSA-N
12598147
C1CC(C2=CC=CC=C2C1)C(=O)O
C11H12O2
MFCD07368364
r-1,2,3,4-tetrahedro-naphthoic acid, r-1,2,3,4-tetrahydronaphthoic acid, r-1,2,3,4-tetrahydro-naphthoic acid, r-1,2,3,4-tetrahydronaphthalene-1-carboxylic acid, r-1,2,3,4-tetrahydro-1-naphthoic acid, r-1,2,3,4-tetrahedronaphthoic acid, 1r-1,2,3,4-tetrahydronaphthalene-1-carboxylic acid, 1-naphthalenecarboxylicacid, 1,2,3,4-tetrahydro-, 1r, pubchem5734, r-1,2,3,4-tert-ahydro naphthoic acid
(1R)-1,2,3,4-tetrahydronaphthalene-1-carboxylic acid

Specifications

23357-47-3
C11H12O2
250 mg
Slightly soluble in water.
+12° (c=1 in Ethanol)
(1R)-1,2,3,4-tetrahydronaphthalene-1-carboxylic acid
12598147
98%
56°C to 58°C
MFCD07368364
r-1,2,3,4-tetrahedro-naphthoic acid, r-1,2,3,4-tetrahydronaphthoic acid, r-1,2,3,4-tetrahydro-naphthoic acid, r-1,2,3,4-tetrahydronaphthalene-1-carboxylic acid, r-1,2,3,4-tetrahydro-1-naphthoic acid, r-1,2,3,4-tetrahedronaphthoic acid, 1r-1,2,3,4-tetrahydronaphthalene-1-carboxylic acid, 1-naphthalenecarboxylicacid, 1,2,3,4-tetrahydro-, 1r, pubchem5734, r-1,2,3,4-tert-ahydro naphthoic acid
VDLWTJCSPSUGOA-SNVBAGLBSA-N
C1CC(C2=CC=CC=C2C1)C(=O)O
176.215
176.22
(R)-1,2,3,4-Tetrahydro-1-naphthoic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only