Learn More
(R)-1,2,3,4-Tetrahydro-1-naphthoic acid, 98%
CAS: 23357-47-3 | C11H12O2 | 176.215 g/mol
Supplier: Thermo Scientific Chemicals H2767306
Description
It is a palonosetron intermediate. Tetrahydronaphthoic acid is used for the construction of sterically constrained protein turn models.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is a palonosetron intermediate. Tetrahydronaphthoic acid is used for the construction of sterically constrained protein turn models.
Solubility
Slightly soluble in water.
Notes
Store in cool dry conditions. Incompatible with oxidizing agents.
Specifications
| (R)-1,2,3,4-Tetrahydro-1-naphthoic acid | |
| 56°C to 58°C | |
| MFCD07368364 | |
| r-1,2,3,4-tetrahedro-naphthoic acid, r-1,2,3,4-tetrahydronaphthoic acid, r-1,2,3,4-tetrahydro-naphthoic acid, r-1,2,3,4-tetrahydronaphthalene-1-carboxylic acid, r-1,2,3,4-tetrahydro-1-naphthoic acid, r-1,2,3,4-tetrahedronaphthoic acid, 1r-1,2,3,4-tetrahydronaphthalene-1-carboxylic acid, 1-naphthalenecarboxylicacid, 1,2,3,4-tetrahydro-, 1r, pubchem5734, r-1,2,3,4-tert-ahydro naphthoic acid | |
| VDLWTJCSPSUGOA-SNVBAGLBSA-N | |
| C1CC(C2=CC=CC=C2C1)C(=O)O | |
| 176.215 | |
| 176.22 |
| 23357-47-3 | |
| C11H12O2 | |
| 5 g | |
| Slightly soluble in water. | |
| +12° (c=1 in Ethanol) | |
| (1R)-1,2,3,4-tetrahydronaphthalene-1-carboxylic acid | |
| 12598147 | |
| 98% |
Chemical Identifiers
| 23357-47-3 | |
| 176.215 | |
| VDLWTJCSPSUGOA-SNVBAGLBSA-N | |
| 12598147 | |
| C1CC(C2=CC=CC=C2C1)C(=O)O |
| C11H12O2 | |
| MFCD07368364 | |
| r-1,2,3,4-tetrahedro-naphthoic acid, r-1,2,3,4-tetrahydronaphthoic acid, r-1,2,3,4-tetrahydro-naphthoic acid, r-1,2,3,4-tetrahydronaphthalene-1-carboxylic acid, r-1,2,3,4-tetrahydro-1-naphthoic acid, r-1,2,3,4-tetrahedronaphthoic acid, 1r-1,2,3,4-tetrahydronaphthalene-1-carboxylic acid, 1-naphthalenecarboxylicacid, 1,2,3,4-tetrahydro-, 1r, pubchem5734, r-1,2,3,4-tert-ahydro naphthoic acid | |
| (1R)-1,2,3,4-tetrahydronaphthalene-1-carboxylic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only