missing translation for 'onlineSavingsMsg'
Learn More

(R)-(+)-1,1,2-Triphenyl-1,2-ethanediol 98.0+%, TCI America™

Catalog No. T14991G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T14991G 1 g
T14995G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T14991G Supplier TCI America Supplier No. T14991G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 95061-46-4
Molecular Formula C20H18O2
Molecular Weight (g/mol) 290.362
MDL Number MFCD00134424
InChI Key GWVWUZJOQHWMFB-LJQANCHMSA-N
PubChem CID 6995538
IUPAC Name (2R)-1,1,2-triphenylethane-1,2-diol
SMILES C1=CC=C(C=C1)C(C(C2=CC=CC=C2)(C3=CC=CC=C3)O)O

Specifications

Melting Point 126°C
Color White
Quantity 1 g
Formula Weight 290.36
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material (R)-(+)-1,1,2-Triphenyl-1,2-ethanediol