missing translation for 'onlineSavingsMsg'
Learn More

Quinoline-3-carboxaldehyde, 98+%

CAS: 13669-42-6 | C10H7NO | 157.172 g/mol

$76.26 - $767.90

Chemical Identifiers

CAS 13669-42-6
Molecular Formula C10H7NO
Molecular Weight (g/mol) 157.172
MDL Number MFCD00006768
InChI Key RYGIHSLRMNXWCN-UHFFFAOYSA-N
Synonym 3-quinolinecarboxaldehyde, quinoline-3-carboxaldehyde, 3-quinolinecarbaldehyde, 3-formylquinoline, quinoline-3-aldehyde, quinoline-3 carbaldehyde, rarechem ak ml 0129, 3-quinoline carboxaldehyde, quinoline 3-carboxaldehyde, 3-quinoline-carboxaldehyde
PubChem CID 83641
IUPAC Name quinoline-3-carbaldehyde
SMILES C1=CC=C2C(=C1)C=C(C=N2)C=O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL13815MD
View Documents
Thermo Scientific Chemicals
L13815MD
250 mg
Each for $76.26
Only null left
 
AAL1381503
View Documents
Thermo Scientific Chemicals
L1381503
1 g
Each for $215.37
Only null left
 
AAL1381506
View Documents
Thermo Scientific Chemicals
L1381506
5 g
Each for $767.90
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

13669-42-6
157.172
RYGIHSLRMNXWCN-UHFFFAOYSA-N
83641
C1=CC=C2C(=C1)C=C(C=N2)C=O
C10H7NO
MFCD00006768
3-quinolinecarboxaldehyde, quinoline-3-carboxaldehyde, 3-quinolinecarbaldehyde, 3-formylquinoline, quinoline-3-aldehyde, quinoline-3 carbaldehyde, rarechem ak ml 0129, 3-quinoline carboxaldehyde, quinoline 3-carboxaldehyde, 3-quinoline-carboxaldehyde
quinoline-3-carbaldehyde

Specifications

13669-42-6
C10H7NO
250 mg
Air Sensitive
RYGIHSLRMNXWCN-UHFFFAOYSA-N
quinoline-3-carbaldehyde
83641
≥98%
68°C to 71°C
MFCD00006768
113269
3-quinolinecarboxaldehyde, quinoline-3-carboxaldehyde, 3-quinolinecarbaldehyde, 3-formylquinoline, quinoline-3-aldehyde, quinoline-3 carbaldehyde, rarechem ak ml 0129, 3-quinoline carboxaldehyde, quinoline 3-carboxaldehyde, 3-quinoline-carboxaldehyde
C1=CC=C2C(=C1)C=C(C=N2)C=O
157.172
157.17
Quinoline-3-carboxaldehyde
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 237-147-7

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only