missing translation for 'onlineSavingsMsg'
Learn More

Quinoline-3-carboxaldehyde, 98+%

CAS: 13669-42-6 | C10H7NO | 157.172 g/mol

Supplier:  Thermo Scientific Chemicals L1381506

 View more versions of this product

Catalog No. AAL1381506


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Quinoline-3-carboxaldehyde
68°C to 71°C
MFCD00006768
113269
3-quinolinecarboxaldehyde, quinoline-3-carboxaldehyde, 3-quinolinecarbaldehyde, 3-formylquinoline, quinoline-3-aldehyde, quinoline-3 carbaldehyde, rarechem ak ml 0129, 3-quinoline carboxaldehyde, quinoline 3-carboxaldehyde, 3-quinoline-carboxaldehyde
C1=CC=C2C(=C1)C=C(C=N2)C=O
157.172
157.17
13669-42-6
C10H7NO
5 g
Air Sensitive
RYGIHSLRMNXWCN-UHFFFAOYSA-N
quinoline-3-carbaldehyde
83641
≥98%

Chemical Identifiers

13669-42-6
157.172
RYGIHSLRMNXWCN-UHFFFAOYSA-N
83641
C1=CC=C2C(=C1)C=C(C=N2)C=O
C10H7NO
MFCD00006768
3-quinolinecarboxaldehyde, quinoline-3-carboxaldehyde, 3-quinolinecarbaldehyde, 3-formylquinoline, quinoline-3-aldehyde, quinoline-3 carbaldehyde, rarechem ak ml 0129, 3-quinoline carboxaldehyde, quinoline 3-carboxaldehyde, 3-quinoline-carboxaldehyde
quinoline-3-carbaldehyde
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 237-147-7

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only