accessibility menu, dialog, popup

UserName

ON

CAS RN 13669-42-6

IUPAC Name: quinoline-3-carbaldehyde
Synonyms: quinoline-3-carbaldehyde
Molecular Weight (g/mol): 157.17
Molecular Formula: C10H7NO
InChi Key: RYGIHSLRMNXWCN-UHFFFAOYSA-N
SMILES: O=CC1=CN=C2C=CC=CC2=C1
Molecular Weight (g/mol) 
  • (2)
  • (5)
Percent Purity 
  • (2)
  • (3)
  • (2)
Quantity 
  • (3)
  • (1)
  • (3)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (2)
  • (5)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (3)
  • (2)
  • (2)
  • (2)