missing translation for 'onlineSavingsMsg'
Learn More

Quinine monohydrochloride dihydrate, 99%

CAS: 6119-47-7 | C20H29ClN2O4 | 396.91 g/mol

$69.53 - $214.45

Chemical Identifiers

CAS 6119-47-7
Molecular Formula C20H29ClN2O4
Molecular Weight (g/mol) 396.91
MDL Number MFCD00151248
InChI Key MPQKYZPYCSTMEI-VQYSEXPNNA-N
Synonym quinine hcl dihydrate
PubChem CID 124080947
IUPAC Name (R)-[(2S,4S,5S)-5-ethenyl-1-azabicyclo[2.2.2]octan-2-yl]-(6-methoxyquinolin-4-yl)methanol;dihydrate;hydrochloride
SMILES O.O.Cl.[H][C@@]1(CN2CCC1C[C@@]2([H])[C@H](O)C1=C2C=C(OC)C=CC2=NC=C1)C=C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC163720050
SDS View Documents
Thermo Scientific Chemicals
163720050
5 g Glass bottle N/A
Only null left
 
AC163720250
SDS View Documents
Thermo Scientific Chemicals
163720250
25 g Glass bottle N/A
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

6119-47-7
396.91
MPQKYZPYCSTMEI-VQYSEXPNNA-N
124080947
O.O.Cl.[H][C@@]1(CN2CCC1C[C@@]2([H])[C@H](O)C1=C2C=C(OC)C=CC2=NC=C1)C=C
C20H29ClN2O4
MFCD00151248
quinine hcl dihydrate
(R)-[(2S,4S,5S)-5-ethenyl-1-azabicyclo[2.2.2]octan-2-yl]-(6-methoxyquinolin-4-yl)methanol;dihydrate;hydrochloride

Specifications

6119-47-7
White
6% to 10% (1 g, 105°C)
99%
C20H29ClN2O4
5 g
15, 8177
− 258.00
Solubility in water: 62.5g/L (20 c). Other solubilities: soluble in alcohol,glycerol and chloroform,slightly soluble in ether
O.O.Cl.[H][C@@]1(CN2CCC1C[C@@]2([H])[C@H](O)C1=C2C=C(OC)C=CC2=NC=C1)C=C
396.91
396.92
Crystalline Powder or Silky Needles
115.0°C to 116.0°C
122°C
Authentic
Glass bottle
MFCD00151248
07,311
− 258.00 (20.00°C c=2,0.1M HCL)
quinine hcl dihydrate
MPQKYZPYCSTMEI-VQYSEXPNNA-N
(R)-[(2S,4S,5S)-5-ethenyl-1-azabicyclo[2.2.2]octan-2-yl]-(6-methoxyquinolin-4-yl)methanol;dihydrate;hydrochloride
124080947
99%
Quinine monohydrochloride dihydrate, 99%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
May cause an allergic skin reaction.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation or rash occurs: Get medical advice/attention.

GHS Signal Word: Warning

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only