Learn More
Quinine monohydrochloride dihydrate, 99%
CAS: 6119-47-7 | C20H29ClN2O4 | 396.91 g/mol
Supplier: Thermo Scientific Chemicals 163720250
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Quinine monohydrochloride dihydrate | |
| 6119-47-7 | |
| White | |
| 6% to 10% (1 g, 105°C) | |
| 99% | |
| C20H29ClN2O4 | |
| 25 g | |
| 15, 8177 | |
| − 258.00 | |
| Solubility in water: 62.5g/L (20 c). Other solubilities: soluble in alcohol,glycerol and chloroform,slightly soluble in ether | |
| O.O.Cl.[H][C@@]1(CN2CCC1C[C@@]2([H])[C@H](O)C1=C2C=C(OC)C=CC2=NC=C1)C=C | |
| 396.91 | |
| 396.92 | |
| Crystalline Powder or Silky Needles |
| 99% | |
| 115.0°C to 116.0°C | |
| 122°C | |
| Authentic | |
| Glass bottle | |
| MFCD00151248 | |
| 07,311 | |
| − 258.00 (20.00°C c=2,0.1M HCL) | |
| quinine hcl dihydrate | |
| MPQKYZPYCSTMEI-VQYSEXPNNA-N | |
| (R)-[(2S,4S,5S)-5-ethenyl-1-azabicyclo[2.2.2]octan-2-yl]-(6-methoxyquinolin-4-yl)methanol;dihydrate;hydrochloride | |
| 124080947 | |
| 99% |
Chemical Identifiers
| 6119-47-7 | |
| 396.91 | |
| MPQKYZPYCSTMEI-VQYSEXPNNA-N | |
| 124080947 | |
| O.O.Cl.[H][C@@]1(CN2CCC1C[C@@]2([H])[C@H](O)C1=C2C=C(OC)C=CC2=NC=C1)C=C |
| C20H29ClN2O4 | |
| MFCD00151248 | |
| quinine hcl dihydrate | |
| (R)-[(2S,4S,5S)-5-ethenyl-1-azabicyclo[2.2.2]octan-2-yl]-(6-methoxyquinolin-4-yl)methanol;dihydrate;hydrochloride |
Safety and Handling
GHS H Statement
Harmful if swallowed.
May cause an allergic skin reaction.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation or rash occurs: Get medical advice/attention.
GHS Signal Word: Warning
RUO – Research Use Only