missing translation for 'onlineSavingsMsg'
Learn More

Quinhydrone, 98%

Catalog No. AC418770250
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC418770250 25 g Glass bottle
AC418771000 100 g Glass bottle
AC418775000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC418770250 Supplier Thermo Scientific Chemicals Supplier No. 418770250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Quinhydrone, 98%, Quantity: 100g, Packaging: Glass bottle, Green, Melting Point: 168 deg.C to 172 deg.C, Molecular Weight: 218.21, Percent Purity: 98%, Assay Percent Range: 48.0 to 52% Hydroquinone (HPLC), CAS: 106-34-3, ChEBI: CHEBI:26491, Density: 1.4000g/mL | CAS: 106-34-3 | C12H10O4 | 218.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 106-34-3
Molecular Formula C12H10O4
Molecular Weight (g/mol) 218.21
MDL Number MFCD00010310
InChI Key BDJXVNRFAQSMAA-UHFFFAOYSA-N
Synonym quinhydrone, green hydroquinone, p-benzoquinhydrone, chinhydron, chinhydron czech, unii-p4a66lq3qj, hydroquinone, compd. with p-benzoquinone, p-benzoquinone, compd. with hydroquinone, 2,5-cyclohexadiene-1,4-dione, compd. with 1,4-benzenediol 1:1, p-benzoquinone-hydroquinone compound 1:1
PubChem CID 7801
ChEBI CHEBI:26491
IUPAC Name benzene-1,4-diol;cyclohexa-2,5-diene-1,4-dione
SMILES OC1=CC=C(O)C=C1.O=C1C=CC(=O)C=C1

Specifications

Melting Point 168°C to 172°C
Color Green
Density 1.4000g/mL
Infrared Spectrum Authentic
Assay Percent Range 48.0 to 52% Hydroquinone (HPLC)
Packaging Glass bottle
Quantity 25 g
Specific Gravity 1.4
Merck Index 15, 8174
Solubility Information Solubility in water: 4g/L (20°C). Other solubilities: soluble in hot water,ammonia,alcohol,ether,insoluble in petroleum ether
Formula Weight 218.21
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material Quinhydrone
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Very toxic to aquatic life.
Toxic if swallowed.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN
EINECSNumber 203-387-6
RTECSNumber VA4550000
TSCA TSCA

RUO – Research Use Only