missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Quinaldine, ≥90% (GC), MilliporeSigma™ Supelco™
$435.86 - $581.71
Specifications
| Packaging | Glass Bottle |
|---|---|
| CAS | 91-63-4 |
| Density | 1.058 g/mL (at 25°C) |
| Flash Point | 79°C |
| Physical Form | Neat |
Description
Pharmaceutical (small molecule)Specifications
| Glass Bottle | |
| 1.058 g/mL (at 25°C) | |
| Neat | |
| MFCD00006756 | |
| 105°C to 107°C (10 mmHg); 248°C | |
| Analytical Standard | |
| ≥90% (GC) |
| 91-63-4 | |
| 79°C | |
| C10H9 N | |
| -9°C to -3°C | |
| 2-Methylquinoline | |
| 143.19 | |
| Analytical Standard |