accessibility menu, dialog, popup

UserName

H3CN

CAS RN 91-63-4

IUPAC Name: 2-methylquinoline
Synonyms: quinaldine
Molecular Weight (g/mol): 143.19
Molecular Formula: C10H9N
InChi Key: SMUQFGGVLNAIOZ-UHFFFAOYSA-N
SMILES: CC1=CC=C2C=CC=CC2=N1
Boiling Point 
  • (3)
  • (3)
  • (2)
Percent Purity 
  • (3)
  • (3)
  • (2)
Quantity 
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
Physical Form 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (3)
  • (5)
  • (3)
  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (3)