missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ QuantStudio™ Absolute Q™ Isolation Buffer

Catalog No. A52730
Change view
Click to view available options
Quantity:
3 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A52730 3 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A52730 Supplier Applied Biosystems™ Supplier No. A52730

missing translation for 'validMainframeMsg'

The QuantStudio Absolute Q Isolation Buffer is intended to supplement the materials supplied in the QuantStudio Absolute Q MAP16 Plate Kit.

  • The QuantStudio Absolute Q Isolation Buffer is one component of the QuantStudio Absolute Q MAP16 Plate Kit (A52865)
  • This buffer is intended to supplement the materials supplied in the plate kit for those customers who require additional materials
  • For use within the QuantStudio Absolute Q Digital PCR System workflow to help partition samples within the reaction wells of the MAP16 plate

Specifications

Detection Method Primer-probe
For Use With (Equipment) QuantStudio™ Absolute Q™ Digital PCR System
Product Line QuantStudio
Quantity 3 mL
Shipping Condition Room Temperature
For Use With (Application) Digital PCR (dPCR)
Compatibility QuantStudio Absolute Q MAP16 Plate Kit
Size (Final Product) 3 mL
I finished running my experiment on the QuantStudio Absolute Q Digital PCR instrument but just realized that I had selected the wrong dyes during setup. Can I change the dyes that were used after the run?

If dye channels are turned off before the run, then data will not be collected for them as the run progresses. This means that changing the dyes after the run will not be possible.

I ran my experiment using the QuantStudio Absolute Q Digital PCR System but did not actually use dyes in all the channels. Is there a way to only see those dye channels that were used?

Unused dye channels can be turned off after the run by going to the SETUP tab and clicking EDIT SETUP and then EDIT GROUPS. Here, the Analysis can be changed to “Not Used.” After clicking SAVE twice, the data will be reanalyzed and the unused dye channels will not be present in the analysis.

I'm using the QuantStudio Absolute Q Digital PCR System and the software is saying that the concentration is NaN. What does that mean?

NaN stands for “not a number”. The software will display NaN instead of an actual number when it has detected some issue during the analysis of the array images.

With the QuantStudio Absolute Q Digital PCR instrument, the run data didn't automatically transfer after the run completed. How can I get the data?

We would recommend restarting the software. If the run still doesn't transfer, we would recommend closing the software, rebooting the instrument, and then starting the software. If further assistance is needed, please contact Technical Support at techsupport@thermofisher.com.

With the QuantStudio Absolute Q Digital PCR System, how much sample can be loaded into each well?

9 µL of sample is loaded into each well along with 15 µL of isolation buffer. Loading more or less of either the sample or isolation buffer will affect filling of the microchambers and the run results.

How do I know how much sample to use with the QuantStudio Absolute Q Digital PCR System?

Please see the sections on determining the optimal dilution in Chapter 2 of the QuantStudio Absolute Q Digital PCR System Installation, Use, and Maintenance Guide (https://assets.thermofisher.com/TFS-Assets/LSG/manuals/MAN0025621_QS_Absolute_Q_Digital_PCR_System_UG_RUO_EN.pdf).

>
How many samples can be run on the QuantStudio Absolute Q Digital PCR instrument at one time?

The QuantStudio Absolute Q MAP16 Digital PCR Plate can accommodate up to 16 individual samples. Loading the same sample in multiple microarrays as replicates or for the purpose of pooling results will decrease the number of individual samples that can be run on the same plate.

How many QuantStudio Absolute Q Digital PCR instruments can be managed at the same time using the QuantStudio Absolute Q Digital PCR Software?

QuantStudio Absolute Q Digital PCR Software can only manage one QuantStudio Absolute Q Digital PCR instrument at a time.

How long does a run on the QuantStudio Absolute Q Digital PCR System take?

A result can be provided in less than 90 mins depending on the protocol that is used.

Are there any special precautions when loading the QuantStudio Absolute Q MAP16 Digital PCR Plate?

Yes. Here are some precautions to take:
- Do not allow the pipette tip to come in contact with the bottom of the well or puncture the thin membrane that resides at the bottom of the well.
- Be careful not to induce bubbles when pipetting into the wells.
- Do not mix the isolation buffer and sample.
- Ensure that the gaskets are oriented the correct way and are pushed all the way on.

For more detailed instructions, please see the guidelines in Chapter 2 of the QuantStudio Absolute Q Digital PCR System Installation, Use, and Maintenance Guide (https://assets.thermofisher.com/TFS-Assets/LSG/manuals/MAN0025621_QS_Absolute_Q_Digital_PCR_System_UG_RUO_EN.pdf).

What master mix can I use with the QuantStudio Absolute Q Digital PCR System?

The QuantStudio Absolute Q Digital PCR System is meant to be used with the Absolute Q DNA Digital PCR Master Mix (5X) (Cat. No. A52490) or Absolute Q 1-step RT-dPCR Master Mix (4X) (Cat. No. A55146). Other master mixes have not been validated with this system.

How do I find the .zst file for the QuantStudio Absolute Q Digital PCR System?

The .zst data files are not automatically generated. To create a .zst file, select the run by checking the box to the left of the run name and then click on the “Export Run” button that appears. You will be asked if that is the run you would like to export and then would need to select a location to export the file to.

What dyes are supported by the QuantStudio Absolute Q Digital PCR System?

The QuantStudio Absolute Q Digital PCR System has filters that support FAM, HEX (not field calibrated), VIC, ABY, ROX (QC channel), CY5, and JUN dyes.

Does the QuantStudio Absolute Q instrument use channels (e.g., ROX dye) to determine which compartments have a reaction?

Yes, ROX dye is the passive reference dye for QC (Quality Control).

Do I need to use the entire QuantStudio Absolute Q MAP16 Digital PCR Plate at one time?

No. Microarrays will be used in columns of 4 and unused columns can be used at a future time.

What does the QuantStudio Absolute Q MAP16 Digital PCR Plate look like?

The QuantStudio Absolute Q MAP16 Digital PCR Plate is a microinjection-molded plate which is 85.5 mm x 128 mm and consists of 16 microarrays. Each microarray consists of approximately 20,000 microchambers, each accommodating 432 pL (picoliter) volume of reaction. See image on page 29 of the QuantStudio Absolute Q Digital PCR System Installation, Use, and Maintenance Guide (https://assets.thermofisher.com/TFS-Assets/LSG/manuals/MAN0025621_QS_Absolute_Q_Digital_PCR_System_UG_RUO_EN.pdf).

What are the electrical and environmental specifications for the QuantStudio Absolute Q Digital PCR System?

For QuantStudio Absolute Q instrument specifications, please see Appendix F of the QuantStudio Absolute Q Digital PCR System Installation, Use, and Maintenance Guide (https://assets.thermofisher.com/TFS-Assets/LSG/manuals/MAN0025621_QS_Absolute_Q_Digital_PCR_System_UG_RUO_EN.pdf).

For the QuantStudio Absolute Q Digital PCR System, what does MAP mean or stand for?

MAP stands for patented Microfluidic Array Plate technology. The QuantStudio Absolute Q Digital PCR instrument uses QuantStudio Absolute Q MAP16 Digital PCR Plates (MAP plates) for loading samples and running experiments. The MAP plates use microinjection-molded plate technology to overcome common reagent distribution challenges of inconsistency and high dead volume. This novel approach provides many benefits, including precise micro-chamber volume, consistent sample digitization, and greater than 95% of the loaded volume analyzed:
- 16 samples per plate
- 20,000 micro-chambers per reaction
- <5% dead volume

For the QuantStudio Absolute Q Digital PCR System, what is the volume per compartment?

Each microchamber holds 432 picoliters (pL).


For Research Use Only. Not for use in diagnostic procedures