missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ QuantStudio™ 3D Digital PCR UV Sealing Kits

Provides the materials needed to ensure an occlusive seal on the QuantStudio™ 3D Digital PCR Chip following reagent addition.

$92.50 - $1,040.00

Specifications

For Use With (Equipment) QuantStudio™
Product Type Sealing Kit
Quantity 1 Kit
View More Specs

Includes:
Contains
  • 1 x QuantStudio™ 3D UV Cure Chip Sealant
  • 1 x QuantStudio™ 3D UV Cure Chip Sealant Tip

Products 2
Catalog Number Mfr. No. Description Content And Storage Price Quantity & Availability  
Catalog Number Mfr. No. Description Content And Storage Price Quantity & Availability  
4485813
Encompass
View Documents Promotion Details
Applied Biosystems™
4485813
With Pen
Contains:
  •  AAAA Batteries for UV-Curing Stylus (6)
  • UV-Activat
Each for $1,040.00
Only null left
 
4488475
Encompass
View Documents Promotion Details
Applied Biosystems™
4488475
Glue
Contains
  • 1 x QuantStudio™ 3D UV Cure Chip Sealant
  • 1 x Q
Each for $92.50
Only null left
 
Description

Description

This is accomplished through the application of sealant and the use of UV light to cure the sealant. This kit includes a syringe of sealant and a sealant tip. After loading sample onto the chip, apply the chip lid, add immersion fluid, and seal the fill-hole with the UV sealant and cure the sealant with a UV light pen. If you do not have a UV pen, an alternate version of this kit is available that includes a UV pen: QuantStudio™ 3D Digital PCR UV Sealing Kit with Pen.

For Research Use Only. Not for use in diagnostic procedures.
Specifications

Specifications

QuantStudio™
Sealing Kit
1 Kit
Videos
SDS
Documents

Documents