missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Applied Biosystems™ QuantStudio™ 3D Digital PCR UV Sealing Kits
Description
Includes
Contains
- 1 x QuantStudio™ 3D UV Cure Chip Sealant
- 1 x QuantStudio™ 3D UV Cure Chip Sealant Tip
This is accomplished through the application of sealant and the use of UV light to cure the sealant. This kit includes a syringe of sealant and a sealant tip. After loading sample onto the chip, apply the chip lid, add immersion fluid, and seal the fill-hole with the UV sealant and cure the sealant with a UV light pen. If you do not have a UV pen, an alternate version of this kit is available that includes a UV pen: QuantStudio™ 3D Digital PCR UV Sealing Kit with Pen.
For Research Use Only. Not for use in diagnostic procedures.
Order Info
Not required for v2 chip lids.
Specifications
Specifications
| Content And Storage | Contains:
|
| For Use With (Equipment) | QuantStudio™ |
| Product Type | Sealing Kit |
| Description | With Pen |
| Quantity | 1 Kit |