missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ QuantStudio™ 3D Digital PCR UV Sealing Kits

Catalog No. 4488475
Encompass
Change view
Click to view available options
Description:
Glue
With Pen
Content And Storage:
Contains
  • 1 x QuantStudio™ 3D UV Cure Chip Sealant
  • 1 x Q
Contains:
  •  AAAA Batteries for UV-Curing Stylus (6)
  • UV-Activat
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Description Content And Storage
4488475 Glue
Contains
  • 1 x QuantStudio™ 3D UV Cure Chip Sealant
  • 1 x Q
4485813 With Pen
Contains:
  •  AAAA Batteries for UV-Curing Stylus (6)
  • UV-Activat
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 4488475 Supplier Applied Biosystems™ Supplier No. 4488475

Includes

Contains
  • 1 x QuantStudio™ 3D UV Cure Chip Sealant
  • 1 x QuantStudio™ 3D UV Cure Chip Sealant Tip

Provides the materials needed to ensure an occlusive seal on the QuantStudio™ 3D Digital PCR Chip following reagent addition.

This is accomplished through the application of sealant and the use of UV light to cure the sealant. This kit includes a syringe of sealant and a sealant tip. After loading sample onto the chip, apply the chip lid, add immersion fluid, and seal the fill-hole with the UV sealant and cure the sealant with a UV light pen. If you do not have a UV pen, an alternate version of this kit is available that includes a UV pen: QuantStudio™ 3D Digital PCR UV Sealing Kit with Pen.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Content And Storage
Contains
  • 1 x QuantStudio™ 3D UV Cure Chip Sealant
  • 1 x Q
For Use With (Equipment) QuantStudio™
Product Type Sealing Kit
Description Glue
Quantity 1 Kit