missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ QuantStudio™ 12K Flex OpenArray™ Vaginal Microbiota Starter Kit

Catalog No. A32087
Encompass
Change view
Click to view available options
Quantity:
1 kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A32087 1 kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A32087 Supplier Applied Biosystems™ Supplier No. A32087

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The QuantStudio™ 12K Flex OpenArray™ Vaginal Microbiota Starter Kit contains two training plates and a synthetic amplification control to serve as a sample for experiments.

  • A variety of starter kits are available to introduce users to the various applications of the QuantStudio™ 12K Flex OpenArray™ sample block
  • Each starter kit contains 2 arrays, plus sufficient reagents and accessories to perform two experiments
  • The QuantStudio™ 12K Flex OpenArray™ Vaginal Microbiota Starter Kit contains two training plates and a synthetic amplification control to serve as a sample for experiments
  • This starter kit serves as a training tool for researchers new to this application

The accessories provided in the kit include: 6 lids and plugs, 6 immersion fluid syringes, 2 carriers, 1 box of Accufill™ System tips, and 384-well plates

Specifications

Detection Method Primer-probe
Format 384-Well Plate
PCR Method qPCR
Product Line TaqMan
Product Type 12K Flex Vaginal Microbiota Starter Kit
Quantity 1 kit
Shipping Condition Dry Ice
Sample Type DNA

For Research Use Only. Not for use in diagnostic procedures.