missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ QuantStudio™ 12K Flex OpenArray™ Calibration Kit

Catalog No. 4478601
Encompass
Change view
Click to view available options
Quantity:
1 kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
4478601 1 kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 4478601 Supplier Applied Biosystems™ Supplier No. 4478601
Season of Savings
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Offered in order to maintain the QuantStudio™ 12K Flex Real Time PCR System OpenArray™ block type.

The calibration kit contains FAM™ dye, as well as a calibration starter kit. The calibration starter kit contains the calibration plaque for background and uniformity, as well as four empty cases for dye loading. Calibration is recommended on a yearly basis. Please see your instrument maintenance guide for full instructions on use.

Specifications

Calibration Type Dye Calibration, Background Calibration, Uniformity Calibration
Content And Storage 1 Bottle Fam Dye, store at -20°C
1 Calibration Starter Kit, store at room temperature
For Use With (Equipment) QuantStudio™ 12k Flex
Label or Dye FAM
Product Line QuantStudio
Product Type PCR Calibration Kit
Quantity 1 kit
What type of Real-Time PCR (Sequence Detection System) information is available from Applied Biosystems?

Tutorials demonstrating introductions to Real-Time PCR, guidelines for designing real-time assays using Primer Express software, and various instrument set-ups can be found under Life Technologies University on our website. User bulletins, manuals and Product inserts, protocols can be found as downloadable pdf files on our Technical Resources page.

Which kit do I need to calibrate the QuantStudio 12K Flex Real-Time PCR System for the OpenArray block?

We recommend performing the calibration for the OpenArray block every 12 months. This can be done using the QuantStudio 12K Flex OpenArray Calibration Kit (Cat. No. 4478601).

For Research Use Only. Not for use in diagnostic procedures.