missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ QuantaBlu™ Fluorogenic Peroxidase Substrate Kit

Catalog No. PI15169
Change view
Click to view available options
Quantity:
530 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI15169 530 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI15169 Supplier Thermo Scientific™ Supplier No. LSG15169
Season of Savings
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Conduct stopped, non-stopped and kinetic assays using this soluble fluorogenic (chemifluorescent) peroxidase substrate for ELISA.

Thermo Scientific™ QuantaBlu Fluorogenic Peroxidase Substrate is a soluble fluorogenic (chemifluorescent) substrate for detection of peroxidase activity in ELISA methods.

Additional Information and Online Ordering

In the presence of active peroxidase the working solution of QuantaBlu Substrate produces a blue fluorescent product that can be quantitated by fluorometry in microplates or cuvettes. Fluorometric-based detection has a large dynamic range, which overcomes the limitations of using spectrophotometry to measure colorimetric substrates. The blue fluorescent product is not sensitive to light and does not photobleach. QuantaBlu Fluorogenic Peroxidase Substrate is stable, highly sensitive, produces high signal-to-noise ratios and allows for a broad dynamic detection range of peroxidase activity. This substrate is also flexible and can be used for stopped, nonstopped and kinetic assays. Incubation for stopped and nonstopped assays can be performed between 5 and 90 minutes at either room temperature (RT) or 37°C. The QuantaBlu NS/K Substrate formulation can be integrated into robotic-based assays and is designed for assays that do not need to be stopped.

Highlights:

  • More sensitive than TMB, OPD or ABTS substrates
  • Flexible stopped, nonstopped or kinetic assays possible
  • Large dynamic range (4 log peroxidase concentration range)
  • Excellent stability; working solution is stable for 24 hours
  • Large stokes shift; excitation/emission maxima of 325nm/420nm (range of 315-345/370-460)
  • Does not photobleach

Specifications

Description QuantaBlu Fluorogenic Peroxidase Substrate
Product Type ELISA Substrate
Quantity 530 mL
Detection Method Fluorescent
Product Line QuantaBlu
Substrate Properties Chemical Substrate
Substrate Type HRP (Horseradish Peroxidase)
Content And Storage Sufficient For: 275 mL of 1X working solution for non-stopped assay (2750 wells at 100 μL per well) or 250 mL of 1X working solution when used 1:1 with stop solution (2500 wells at 100 μL per well)
• QuantaBlu Substrate Solution, 250 mL
• QuantaBlu Peroxide Solution, 30 mL
• QuantaBlu Stop Solution, 250 mL
Store at 4°C.

For Research Use Only. Not for use in diagnostic procedures.