missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Qorpak™ White Metal Pulp/Aluminum Foil Lined Caps
Free of odor and taste problems
$1220.31 - $7160.34
Specifications
| Color | White |
|---|---|
| Material | Metal |
| Liner Material | Pulp/Aluminum Foil |
| For Use With (Equipment) | Wide mouth bottle |
| Catalog Number | Mfr. No. | Neck Finish | No. per Case | Packaging | Price | Quantity | |||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Neck Finish | No. per Case | Packaging | Price | Quantity | |||
|
36-101-0824
|
Qorpak™
CAP00451 |
58-400 | 2000 Cs. | Bulk |
Case of 2500 Each for $5,748.26
|
|
|||
|
02-991-189
|
Qorpak™
CAP00434 |
28-400 | 5400 Cs. | Bulk |
Case of 5400 Each for $7,160.34
|
|
|||
|
02-993-384
|
Qorpak™
CAP06647 |
28-400 | 576 Cs. | Bagged |
Case of 576 Each for $1,220.31
|
|
|||
Description
- Made of steel coated with anti-corrosive tinplate
- Impact resistant
- Tolerate wide ranges of temperature
- Feature standard 400 GPI thread finish
- Used for non-acid, non-alkaline products, organic solvents, chrome cleaners, food bottles and jars, brake fluids and mineral oils
Specifications
| White | |
| Metal | |
| Pulp/Aluminum Foil | |
| Wide mouth bottle |