Learn More
Qorpak™ White Metal Pulp/Aluminum Foil Lined Caps
Free of odor and taste problems
Supplier: Qorpak™ CAP00451
Description
- Made of steel coated with anti-corrosive tinplate
- Impact resistant
- Tolerate wide ranges of temperature
- Feature standard 400 GPI thread finish
- Used for non-acid, non-alkaline products, organic solvents, chrome cleaners, food bottles and jars, brake fluids and mineral oils
Specifications
| White | |
| 2000 Cs. | |
| Pulp/Aluminum Foil | |
| Wide mouth bottle |
| Metal | |
| Bulk | |
| 58-400 |