Learn More
Qorpak™ White Metal Pulp/Aluminum Foil Lined Caps
Free of odor and taste problems
Supplier:  Qorpak™ CAP06647
 View more versions of this product
Catalog No. 02-993-384
Only null left
Explore available promotions
Description
Description
  • Made of steel coated with anti-corrosive tinplate
  • Impact resistant
  • Tolerate wide ranges of temperature
  • Feature standard 400 GPI thread finish
  • Used for non-acid, non-alkaline products, organic solvents, chrome cleaners, food bottles and jars, brake fluids and mineral oils
Specifications
Specifications
White
576 Cs.
Bagged
28-400
Metal
12/Pk.
Pulp/Aluminum Foil
Wide mouth bottle
Product Suggestions
Product Suggestions