missing translation for 'onlineSavingsMsg'
Learn More

Qorpak™ Clear Glass Jugs with Green Thermoset F217 and PTFE Disc

Storage solution for large quantities of liquid.

$124.09

Specifications

Color Clear
Includes Molded finger grip
Material Glass
Closure Color Green
Closure Liner F217, PTFE
View More Specs

Products 1
Catalog Number Mfr. No. Capacity (Metric) Packaging Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Capacity (Metric) Packaging Quantity Price Quantity & Availability  
02991896
View Documents Promotion Details
Qorpak™
GLC01429
3785 mL Qorpak Carton 4/Cs.
Case of 4 Each for $124.09
Only null left
 
Description

Description

  • Features molded finger grip
  • With green thermoset F217 and PTFE disc
Specifications

Specifications

Clear
Glass
F217, PTFE
38-400 mm
Molded finger grip
Green
Thermoset
Screw Cap
Videos
SDS
Documents

Documents