missing translation for 'onlineSavingsMsg'
Learn More

Qorpak™ Clear Glass Jugs with Black Phenolic Pulp/Vinyl Cap

Storage solution for large quantities of liquid.

$135.29

Specifications

Color Clear
Includes Molded finger grip
Material Glass
Closure Color Black
Closure Liner Pulp/Vinyl
View More Specs

Products 1
Catalog Number Mfr. No. Capacity (Metric) Packaging Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Capacity (Metric) Packaging Quantity Price Quantity & Availability  
033272
View Documents Promotion Details
Qorpak™
GLC01420
1892.71 mL Plain Carton 6/Cs.
Case of 6 Each for $135.29
Only null left
 
Description

Description

  • Molded finger grip
  • Black phenolic pulp/vinyl cap
Specifications

Specifications

Clear
Glass
Pulp/Vinyl
38-400 mm
Molded finger grip
Black
Phenolic
Screw Cap
Videos
SDS
Documents

Documents