Learn More
QIAGEN QIAcard FTA Indicating Micro
For collection, stabilization, processing, transport and archiving of buccal nucleic acid samples. Indicating QIAcard FTA Micro with 1 sample area per card.
$459.98 - $1216.70
Specifications
Description Collect, stabilize, process, transport, and archive buccal DNA samples with Indicating FTA Micro Cards. With FTA, DNA stays in the matrix.
Sample Type Buccal Cells, Saliva
For Use With (Application) Forensics, Transgenic identification, Transfusion medicine/HLA typing, Plasmid screening, Food and agriculture testing, Drug discovery, Genomics, STR analysis, Animal identification, Whole genome amplification, Molecular biology
No. of Samples 1
Product Type Indicating FTA Micro Card
View More Specs
Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
09-800-993
View Documents
QIAGEN
WB120311
25 Pk.
Pack of 25 Each for $459.98
Only null left
09-923-345
View Documents
QIAGEN
WB120211
100 Pk.
Pack of 100 Each for $1,216.70
Only null left
Description
Description
For collection, stabilization, processing, transport and archiving of buccal nucleic acid samples. Indicating QIAcard FTA Micro with 1 sample area per card.
  • QIAcard Indicating FTA Cards include a pink dye that turns white when a colorless sample, e.g. buccal cells or saliva is applied.
  • Stabilization and protection of nucleic acids at room temperature with for spot volumes of 125 μL.
  • Cell lysis on contact, denatures proteins and protects DNA from degradation.
Specifications
Specifications
Collect, stabilize, process, transport, and archive buccal DNA samples with Indicating FTA Micro Cards. With FTA, DNA stays in the matrix.
Forensics, Transgenic identification, Transfusion medicine/HLA typing, Plasmid screening, Food and agriculture testing, Drug discovery, Genomics, STR analysis, Animal identification, Whole genome amplification, Molecular biology
Indicating FTA Micro Card
Buccal Cells, Saliva
1
125 μL
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'