missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Q-BioTaq™ Core Kit 10

A thermostable DNA polymerase for multiplex PCR and RAPDs.

Supplier:  MP Biomedicals™ 11EPQBK1025

Catalog No. 11EPQBK1025

Only null left
Explore available promotions

Includes: Q-BioTaq DNA polymerase, high purity dNTPs Mix (>99%) at 10mM each, 10X Q-BioTaq Buffer with Mg, 10X Q-BioTaq Buffer without Mg, 25mM MgCl2 solution

Description

Description

Q-BioTaq DNA Polymerase is a thermostable DNA polymerase which can be used effectively for PCR and DNA sequencing by the chain termination method. This enzyme is encoded by a modified form of the Thermus aquaticus DNA polymerase gene with an N-terminal deletion. The properties of Q-BioTaq DNA Polymerase include high thermostability (98°C) and absence of 5'-3' exonuclease activity. This polymerase is particularly useful in applications where multiple amplifications are required. The enzyme is highly purified and is free of endo- and exonucleases.

Unit Definition: One unit is the amount of enzyme required to catalyse the incorporation of 10nmol of nucleotides into acid insoluble material in 30 min. at 74 °C under assay conditions.

Certifications

Free of RNases and endo- and exonucleases

Specifications

Specifications

5 U/μL
Q-BioTaq DNA polymerase
Standard PCR
Q-BioTaq DNA polymerase, high purity dNTPs Mix (>99%) at 10mM each, 10X Q-BioTaq Buffer with Mg, 10X Q-BioTaq Buffer without Mg, 25mM MgCl2 solution. Store at -20°C
250U
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

Research Use Only (RUO).