missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pyruvic Acid, 95-100%, Spectrum™ Chemical
Specifications
Specifications
| CAS Max % | 1 |
| Boiling Point | 165°C |
| Packaging | Amber Glass Bottle |
| Quantity | 5 kg |
| Percent Purity | 95 to 100% |
| Grade | Reagent |
| Physical Form | Liquid |
CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.