missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FUJIFILM Wako PYROSTAR™ ES-F Single Test
Endotoxin-specific and specially formulated to be unreactive to (1,3)-ß-D-glucan, Includes pre-dispensed LAL reagent for single measurement, for use with Toxinometer™ and to assist labs transitioning from gel clot to KTA methodology
$624.29 - $870.07
Specifications
| Form | Lyophilized |
|---|---|
| Fill Volume | 200 μL |
| Includes | 25 single test vials and 1 vial of CSE (500 ng/vial) |
| Quantity | 25 Tests |
| Catalog Number | Mfr. No. | Sensitivity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Sensitivity | Price | Quantity & Availability | |||||
|
12333010
|
FUJIFILM Wako
WPESK0015 |
0.015 EU/mL |
Each for $870.07
|
|
|||||
|
12333011
|
FUJIFILM Wako
WPESK0003 |
0.03 EU/mL |
Each for $870.07
|
|
|||||
|
12333012
|
FUJIFILM Wako
WPESK0125 |
0.125 EU/mL |
Each for $624.29
|
|
|||||
|
12333013
|
FUJIFILM Wako
WPESK0025 |
0.25 EU/mL |
Each for $870.07
|
|
|||||
Description
- Endotoxin-specific reagent avoids false positive results from glucans
- KTA sensitivities range from 0.001 to 10 EU/mL
- Can be used with the traditional water bath or heat block
- Can be used with the Toxinometer™ to facilitate easier transition from gel clot to KTA
Specifications
| Lyophilized | |
| 200 μL | |
| 25 single test vials and 1 vial of CSE (500 ng/vial) | |
| 25 Tests |