missing translation for 'onlineSavingsMsg'
Learn More

FUJIFILM Wako PYROSTAR™ ES-F Single Test

Catalog No. 12333010
Change view
Click to view available options
Sensitivity:
0.015 EU/mL
0.03 EU/mL
0.125 EU/mL
0.25 EU/mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sensitivity
12333010 0.015 EU/mL
12333011 0.03 EU/mL
12333012 0.125 EU/mL
12333013 0.25 EU/mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 12333010 Supplier FUJIFILM Wako Supplier No. WPESK0015

Endotoxin-specific and specially formulated to be unreactive to (1,3)-ß-D-glucan, Includes pre-dispensed LAL reagent for single measurement, for use with Toxinometer™ and to assist labs transitioning from gel clot to KTA methodology

  • Endotoxin-specific reagent avoids false positive results from glucans
  • KTA sensitivities range from 0.001 to 10 EU/mL
  • Can be used with the traditional water bath or heat block
  • Can be used with the Toxinometer™ to facilitate easier transition from gel clot to KTA

Specifications

Form Lyophilized
Fill Volume 200 μL
Includes 25 single test vials and 1 vial of CSE (500 ng/vial)
Quantity 25 Tests
Sensitivity 0.015 EU/mL