missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FUJIFILM Wako PYROSTAR™ ES-F Single Test
Description
- Endotoxin-specific reagent avoids false positive results from glucans
- KTA sensitivities range from 0.001 to 10 EU/mL
- Can be used with the traditional water bath or heat block
- Can be used with the Toxinometer™ to facilitate easier transition from gel clot to KTA
Specifications
Specifications
| Form | Lyophilized |
| Fill Volume | 200 μL |
| Includes | 25 single test vials and 1 vial of CSE (500 ng/vial) |
| Quantity | 25 Tests |
| Sensitivity | 0.125 EU/mL |