missing translation for 'onlineSavingsMsg'
Learn More

FUJIFILM Wako PYROSTAR™ ES-F 2 mL Bulk Kit

Endotoxin-specific and specially formulated to be unreactive to (1,3)-ß-D-glucan, Dual-purpose formulated to be used as gel clot or kinetic turbidimetric assay, Bulk configuration for large volume labs

$12,572.21

Specifications

Form Lyophilized
Fill Volume 2 mL
Includes 100 x 2 mL vials of lysate
Quantity 100 Vials
View More Specs

Products 5
Catalog Number Mfr. No. Sensitivity Price Quantity & Availability  
Catalog Number Mfr. No. Sensitivity Price Quantity & Availability  
12333014
View Documents Promotion Details
FUJIFILM Wako
WPEM20015
0.015 EU/mL N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
12333015
View Documents Promotion Details
FUJIFILM Wako
WPEM20003
0.03 EU/mL N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
12333016
View Documents Promotion Details
FUJIFILM Wako
WPEM20006
0.06 EU/mL N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
12333017
View Documents Promotion Details
FUJIFILM Wako
WPEM20125
0.125 EU/mL N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
12333018
View Documents Promotion Details
FUJIFILM Wako
WPEM20025
0.25 EU/mL N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

  • Endotoxin-specific reagent avoids false positive results from glucans
  • Dual-purpose reagent can perform either gel clot or kinetic turbidimetric assay (KTA)
  • Kinetic turbidimetric sensitivities range from 0.001 to 10 EU/mL
  • Due to high sensitivity, chances of interference are reduced
  • Facilitates easier transition from gel clot to KTA
Specifications

Specifications

Lyophilized
2 mL
100 x 2 mL vials of lysate
100 Vials
Videos
SDS
Documents

Documents