Learn More
FUJIFILM Wako PYROSTAR™ ES-F 2 mL Bulk Kit
Endotoxin-specific and specially formulated to be unreactive to (1,3)-ß-D-glucan, Dual-purpose formulated to be used as gel clot or kinetic turbidimetric assay, Bulk configuration for large volume labs
Supplier:  FUJIFILM Wako WPEM20006
 View more versions of this product
Catalog No. 12333016
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description
Description
  • Endotoxin-specific reagent avoids false positive results from glucans
  • Dual-purpose reagent can perform either gel clot or kinetic turbidimetric assay (KTA)
  • Kinetic turbidimetric sensitivities range from 0.001 to 10 EU/mL
  • Due to high sensitivity, chances of interference are reduced
  • Facilitates easier transition from gel clot to KTA
Specifications
Chemical Identifiers
Lyophilized
2 mL
100 x 2 mL vials of lysate
100 Vials
Specifications
Lyophilized
100 x 2 mL vials of lysate
0.06 EU/mL
2 mL
100 Vials
Product Suggestions
Product Suggestions