missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Pyrocatechol Violet, pure, indicator grade

CAS: 115-41-3 | C19H14O7S | 386.37 g/mol

$140.70 - $470.78

Chemical Identifiers

CAS 115-41-3
Molecular Formula C19H14O7S
Molecular Weight (g/mol) 386.37
MDL Number MFCD00005868
InChI Key RRRCKIRSVQAAAS-UHFFFAOYSA-N
Synonym Pyrocatecholsulfonephthalein
PubChem CID 66993
IUPAC Name 4-[3-(3,4-dihydroxyphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]benzene-1,2-diol
SMILES OC1=CC=C(C=C1O)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC=C(O)C(O)=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC146540050
SDS View Documents
Thermo Scientific Chemicals
146540050
5 g
Each for $140.70
Only null left
 
AC146540250
SDS View Documents
Thermo Scientific Chemicals
146540250
25 g
Each for $470.78
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
Specifications

Chemical Identifiers

115-41-3
386.37
RRRCKIRSVQAAAS-UHFFFAOYSA-N
66993
OC1=CC=C(C=C1O)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC=C(O)C(O)=C1
C19H14O7S
MFCD00005868
Pyrocatecholsulfonephthalein
4-[3-(3,4-dihydroxyphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]benzene-1,2-diol

Specifications

115-41-3
MFCD00005868
Solubility in water: soluble. Other solubilities: soluble in ethanol, 2-methoxyethanol
Authentic
4-[3-(3,4-dihydroxyphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]benzene-1,2-diol
66993
386.36
Glass bottle
for complexometric det. of metal oxides: passes
5 g
Pyrocatechol Violet
C19H14O7S
Pyrocatecholsulfonephthalein
RRRCKIRSVQAAAS-UHFFFAOYSA-N
OC1=CC=C(C=C1O)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC=C(O)C(O)=C1
386.37
441 to 449nm (in methanol)
Pure
375 (at lambda max, methanol) min.
Brown or Green
Metallic or Crystalline Powder
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only