missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Pyrocatechol Violet, pure, indicator grade

Catalog No. ac146540050
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC146540050 5 g
AC146540250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC146540050 Supplier Thermo Scientific Chemicals Supplier No. 146540050
Only null left

CAS: 115-41-3 | C19H14O7S | 386.37 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator

Chemical Identifiers

CAS 115-41-3
Molecular Formula C19H14O7S
Molecular Weight (g/mol) 386.37
MDL Number MFCD00005868
InChI Key RRRCKIRSVQAAAS-UHFFFAOYSA-N
Synonym Pyrocatecholsulfonephthalein
PubChem CID 66993
IUPAC Name 4-[3-(3,4-dihydroxyphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]benzene-1,2-diol
SMILES OC1=CC=C(C=C1O)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC=C(O)C(O)=C1

Specifications

Solubility Information Solubility in water: soluble. Other solubilities: soluble in ethanol, 2-methoxyethanol
Infrared Spectrum Authentic
Lambda Max 441 to 449nm (in methanol)
Formula Weight 386.36
Grade Pure
Packaging Glass bottle
Specific Extinction 375 (at lambda max, methanol) min.
Suitability for complexometric det. of metal oxides: passes
Color Brown or Green
Quantity 5 g
Physical Form Metallic or Crystalline Powder
Chemical Name or Material Pyrocatechol Violet
Show More Show Less

RUO – Research Use Only