missing translation for 'onlineSavingsMsg'
Learn More

Pyrilamine Maleate 98.0+%, TCI America™

Supplier:  TCI America P236925G

Quantity 25 g

 View more versions of this product

Catalog No. P236925G


Only null left

Description

Description

Specifications

Chemical Identifiers

59-33-6
401.46
JXYWFNAQESKDNC-BTJKTKAUSA-N
5284451
OC(=O)\C=C/C(O)=O.COC1=CC=C(CN(CCN(C)C)C2=CC=CC=N2)C=C1
C21H27N3O5
MFCD00069333
pyrilamine maleate, mepyramine maleate, anisopyradamine, histosol, minihist, diaminide maleate, paraminyl maleate, antihist, histatex, histine
(2Z)-but-2-enedioic acid; N-[2-(dimethylamino)ethyl]-N-[(4-methoxyphenyl)methyl]pyridin-2-amine

Specifications

Pyrilamine Maleate
102°C
C21H27N3O5
MFCD00069333
JXYWFNAQESKDNC-BTJKTKAUSA-N
(2Z)-but-2-enedioic acid; N-[2-(dimethylamino)ethyl]-N-[(4-methoxyphenyl)methyl]pyridin-2-amine
5284451
≥98.0% (HPLC,T)
59-33-6
White
25 g
pyrilamine maleate, mepyramine maleate, anisopyradamine, histosol, minihist, diaminide maleate, paraminyl maleate, antihist, histatex, histine
OC(=O)\C=C/C(O)=O.COC1=CC=C(CN(CCN(C)C)C2=CC=CC=N2)C=C1
401.46
401.46
Crystalline Powder
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : UT1225000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions